1-Chlor-2,4-dinitrobenzen ≥99.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

C0162-25G C0162-500G
TCIAC0162-25GEA 132 DKK
TCIAC0162-25G TCIAC0162-500G
1-Chlor-2,4-dinitrobenzen ≥99.0% (by GC)
1-Chlor-2,4-dinitrobenzen
Formel: C₆H₃ClN₂O₄
Kogepunkt: 315 °C (1013 hPa)
Smeltepunkt: 52…54 °C
Massefylde: 1,7 g/cm³ (20 °C)
Flammepunkt: 194 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007075
CAS-nummer: 97-00-7
EINECS: 202-551-4
UN nummer: 3441
ADR: 6.1,II
Merck Index: 14,02136
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b253ee6d2df1b9db1cadd3d1984dc040d59b7b6a&vid=e23c6406773677e50d10db44b2ad4b74cafc69bf&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Green powder to crystal
Purity (GC) min. 99.0 %
Melting point 50.0 to 54.0 deg-C
Solubility in EtOH (95vol%) almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR