2,5-Dichlor-4-nitroanilin ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
D2722-25G
D2722-5G
TCIAD2722-25GEA
1740
DKK
TCIAD2722-25G
TCIAD2722-5G
2,5-Dichlor-4-nitroanilin ≥98.0% (by GC)
2,5-Dichlor-4-nitroanilin
Formel:
C₆H₄Cl₂N₂O₂ Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00025157 CAS-nummer: 6627-34-5 EINECS: 229-591-5 ADR: 6.1,III |
|
Specifikationer Testresultater
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 154.0 to 157.0 deg-C |
Solubility in Methanol | faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...