3-Chloranilin ≥99.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
C0110-25ML
C0110-500ML
TCIAC0110-25MLEA
132
DKK
TCIAC0110-25ML
TCIAC0110-500ML
3-Chloranilin ≥99.0% (by GC)
3-Chloranilin
Formel:
C₆H₆ClN Kogepunkt: 230°C Smeltepunkt: –10°C Massefylde: 1,216 g/cm³ (20 °C) Flammepunkt: 118°C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00007765 CAS-nummer: 108-42-9 EINECS: 203-581-0 UN nummer: 2019 ADR: 6.1,II Merck Index: 14,02118 |
|
Specifikationer Testresultater
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...