2,4,6-Trichloranilin ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
T0375-25G
T0375-500G
TCIAT0375-25GEA
156
DKK
TCIAT0375-25G
TCIAT0375-500G
2,4,6-Trichloranilin ≥98.0% (by GC)
2,4,6-Trichloranilin
Formel:
C₆H₄Cl₃N Kogepunkt: 262 °C (1013 hPa) Smeltepunkt: 74 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00007663 CAS-nummer: 634-93-5 EINECS: 211-219-8 UN nummer: 3077 ADR: 9,III |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 77.0 to 81.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...