4-Chlor-2-iodanilin ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

C1800-25G C1800-5G
TCIAC1800-25GEA 1890 DKK
TCIAC1800-25G TCIAC1800-5G
4-Chlor-2-iodanilin ≥98.0% (by GC)
4-Chlor-2-iodanilin
Formel: C₆H₅ClIN
Molekylvægt: 253.47 g/mol
MDL Number: MFCD01863737
CAS-nummer: 63069-48-7
EINECS: 263-839-3
UN nummer: 2811
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cbaa7222e9aec57a3c8d1c4954a4110a4f2cbae5&vid=510f92717df7fcb5f04f98d8714d9c7b9d5c53d3&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Purity (GC) min. 98.0 %
Melting point gt. 40.0 to 44.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR