4-Chlor-3-fluorphenol ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
C2476-25G
C2476-5G
TCIAC2476-25GEA
2220
DKK
TCIAC2476-25G
TCIAC2476-5G
4-Chlor-3-fluorphenol ≥98.0% (by GC, titration analysis)
4-Chlor-3-fluorphenol
Formel:
C₆H₄ClFO Kogepunkt: 84°C Smeltepunkt: 49…53 °C |
MDL Number:
MFCD00042583 CAS-nummer: 348-60-7 ADR: 8,III |
|
Specifikationer Testresultater
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 52.0 to 56.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...