5-Chlor-2-Fluoranilin ≥97.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
C1596-25G
C1596-5G
TCIAC1596-25GEA
903
DKK
TCIAC1596-25G
TCIAC1596-5G
5-Chlor-2-Fluoranilin ≥97.0% (by GC)
5-Chlor-2-Fluoranilin
Formel:
C₆H₅ClFN Kogepunkt: 105°C Flammepunkt: 103°C |
MDL Number:
MFCD00069416 CAS-nummer: 2106-05-0 EINECS: 218-284-1 ADR: 6.1,III |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...