3-Chlor-2-Fluoranilin ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
C1659-25G
C1659-5G
TCIAC1659-25GEA
517
DKK
TCIAC1659-25G
TCIAC1659-5G
3-Chlor-2-Fluoranilin ≥98.0% (by GC)
3-Chlor-2-Fluoranilin
Formel:
C₆H₅ClFN Kogepunkt: 99°C |
MDL Number:
MFCD00069415 CAS-nummer: 2106-04-9 EINECS: 218-283-6 ADR: 6.1,III |
|
Specifikationer Testresultater
Appearance | Colorless to Brown clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...