2-Chlor-6-Fluoranilin ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

C2126-1G C2126-5G
TCIAC2126-1GEA 238 DKK
TCIAC2126-1G TCIAC2126-5G
2-Chlor-6-Fluoranilin ≥98.0% (by GC)
2-Chlor-6-Fluoranilin
Formel: C₆H₅ClFN
Molekylvægt: 145.56 g/mol
Kogepunkt: 91°C
Smeltepunkt: 32°C
Massefylde: 1.349
MDL Number: MFCD00040309
CAS-nummer: 363-51-9
EINECS: 206-657-1

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR