3-Chlor-4-fluorbenzaldehyd ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
C1498-25G
C1498-5G
TCIAC1498-25GEA
1010
DKK
TCIAC1498-25G
TCIAC1498-5G
3-Chlor-4-fluorbenzaldehyd ≥98.0% (by GC)
3-Chloro-4-fluorobenzaldehyde
Formel:
C₇H₄ClFO Kogepunkt: 66 °C (2 -rr) Smeltepunkt: 44…46 °C |
MDL Number:
MFCD00011735 CAS-nummer: 34328-61-5 |
|
Specifikationer Testresultater
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Freezing point | 22.0 to 25.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...