2-Amino-5-chlorbenzoesyre ≥99.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Amino-5-chlorobenzoic acid

A0665-25G A0665-5G
TCIAA0665-25GEA 485 DKK
TCIAA0665-25G TCIAA0665-5G
2-Amino-5-chlorbenzoesyre ≥99.0% (by titrimetric analysis)
2-Amino-5-chlorbenzoesyre
Formel: C₇H₆ClNO₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007838
CAS-nummer: 635-21-2
EINECS: 211-230-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2bb8dc245a78618e29924bb9e87930002e650c3&vid=c71b3fb0b7240763d6a3ac4f0fcc20ad3ac34a1b&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 99.0 %
Melting point 207.0 to 212.0 deg-C
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR