4-Chlorbenzonitril ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
C0135-25G
C0135-500G
TCIAC0135-25GEA
189
DKK
TCIAC0135-25G
TCIAC0135-500G
4-Chlorbenzonitril ≥98.0% (by GC)
4-Chlorbenzonitril
Formel:
C₇H₄ClN Kogepunkt: 223°C Smeltepunkt: 90…93 °C Massefylde: 1,1133 g/cm³ (16.8 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00001813 CAS-nummer: 623-03-0 EINECS: 210-765-4 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 91.0 to 94.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...