2-Chlorbenzonitril ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

C1130-25G C1130-500G
TCIAC1130-25GEA 132 DKK
TCIAC1130-25G TCIAC1130-500G
2-Chlorbenzonitril ≥98.0% (by GC)
2-Chlorbenzonitril
Formel: C₇H₄ClN
Kogepunkt: 232°C
Smeltepunkt: 41…44 °C
Massefylde: 1,19 g/cm³ (25 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00001779
CAS-nummer: 873-32-5
EINECS: 212-836-5
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a973a7836b267318a0d46da79369e1e31f679b7&vid=316d5143224004d4f17dc837955e9bf2aa0e958d&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 43.0 to 46.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR