2-Chlorveratraldehyd ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

C1997-25G C1997-5G
TCIAC1997-25GEA 673 DKK
TCIAC1997-25G TCIAC1997-5G
2-Chlorveratraldehyd ≥98.0% (by GC)
2-Chlorveratraldehyd
Formel: C₉H₉ClO₃
MDL Number: MFCD00084942
CAS-nummer: 5417-17-4
EINECS: 226-515-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2181afcf1c852311c03bbaa819b2c9922504d6d6&vid=d258348896bcaa2eb5ee7df4c6328b4ae4fb4eac&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 69.0 to 73.0 deg-C
Solubility in Toluene almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR