3-Phenoxybenzoesyre ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
P1253-25G
P1253-5G
TCIAP1253-25GEA
838
DKK
TCIAP1253-25G
TCIAP1253-5G
3-Phenoxybenzoesyre ≥98.0% (by GC, titration analysis)
3-Phenoxybenzoesyre
Formel:
C₁₃H₁₀O₃ Molekylvægt: 214.22 g/mol Kogepunkt: 365 Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00002498 CAS-nummer: 3739-38-6 EINECS: 223-121-2 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 145.0 to 149.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...