2-Phenoxybenzoesyre

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

P1252-25G P1252-5G
TCIAP1252-25GEA 1510 DKK
TCIAP1252-25G TCIAP1252-5G
2-Phenoxybenzoesyre
2-Phenoxybenzoesyre
Formel: C₁₃H₁₀O₃
Molekylvægt: 214.22 g/mol
Kogepunkt: 355°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00002429
CAS-nummer: 2243-42-7
EINECS: 218-811-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=43f5c696c192c8f621b482a6146ba38194cc4456&vid=f61e6f52d956c16966c54b78e62d0f91e521ae9b&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 99.0 %
Melting point 112.0 to 115.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR