4-Methoxybenzoesyre ≥99.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 4-Methoxybenzoic acid, 4-Anisic acid

A0482-500G A0482-25G
TCIAA0482-500GEA 591 DKK
TCIAA0482-500G TCIAA0482-25G
4-Methoxybenzoesyre ≥99.0% (by GC, titration analysis)
4-Methoxybenzoesyre
Formel: C₈H₈O₃
Kogepunkt: 275 °C (1013 hPa)
Smeltepunkt: 182…185 °C
Massefylde: ~ 1,385 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00002542
CAS-nummer: 100-09-4
EINECS: 202-818-5
Merck Index: 14,00666
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=89195d9f48e5a310fc4a0850238230d78b334dd9&vid=8063fb679f508b4b4323a70d4cd4bb1f93ccb5aa&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Purity (Neutralization titration) min. 99.0 %
Melting point 181.0 to 186.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR