4-Amino-2-hydroxybenzoesyre ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
4-Amino-2-hydroxybenzoic acid
A0420-25G
A0420-100G
TCIAA0420-25GEA
427
DKK
TCIAA0420-25G
TCIAA0420-100G
4-Amino-2-hydroxybenzoesyre ≥98.0% (by titrimetric analysis)
4-Amino-2-hydroxybenzoesyre
Formel:
C₇H₇NO₃ Molekylvægt: 153.14 g/mol Kogepunkt: 347 Smeltepunkt: 150 Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00007789 CAS-nummer: 65-49-6 EINECS: 200-613-5 Merck Index: 14,00477 |
|
Specifikationer Testresultater
Appearance | White to Light yellow to Dark green powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...