3-Aminosalicylsyre ≥98.0% (by HPLC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
3-Aminosalicylic Acid, 3-Amino-2-hydroxybenzoic acid, 3-Amino-2-hydroxybenzoic acid
A0421-1G
A0421-5G
TCIAA0421-1GEA
403
DKK
TCIAA0421-1G
TCIAA0421-5G
3-Aminosalicylsyre ≥98.0% (by HPLC)
3-Aminosalicylsyre
Formel:
C₇H₇NO₃ Kogepunkt: 347 °C (1013 hPa) Smeltepunkt: 290 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00010299 CAS-nummer: 570-23-0 EINECS: 209-328-0 |
|
Specifikationer Testresultater
Appearance | Light yellow to Brown powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 97.0 % |
Solubility in 1mol/L NaOH | within almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...