4-Amino-3-nitrobenzoesyre ≥95.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
A1417-25G
A1417-500G
A1417-100G
TCIAA1417-25GEA
600
DKK
TCIAA1417-25G
TCIAA1417-500G
TCIAA1417-100G
4-Amino-3-nitrobenzoesyre ≥95.0% (by HPLC, titration analysis)
4-Amino-3-nitrobenzoesyre
Formel:
C₇H₆N₂O₄ Smeltepunkt: 290 °C |
MDL Number:
MFCD00017009 CAS-nummer: 1588-83-6 EINECS: 216-453-4 |
|
Specifikationer Testresultater
Appearance | White to Amber powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Purity (Neutralization titration) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...