Pentylbenzoat ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

B1095-25ML B1095-500ML B1095-100ML
TCIAB1095-25MLEA 271 DKK
TCIAB1095-25ML TCIAB1095-500ML TCIAB1095-100ML
Pentylbenzoat ≥98.0% (by GC)
Pentylbenzoat

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR