Isoamylbenzoat ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Isoamyl Benzoate, Benzoic acid isopentyl ester
B0071-25G
B0071-500G
TCIAB0071-25GEA
132
DKK
TCIAB0071-25G
TCIAB0071-500G
Isoamylbenzoat ≥98.0% (by GC)
Isoamylbenzoat
Formel:
C₁₂H₁₆O₂ Molekylvægt: 192.26 g/mol Kogepunkt: 262°C Flammepunkt: 129°C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00026515 CAS-nummer: 94-46-2 EINECS: 202-334-4 Merck Index: 14,05113 |
|
Specifikationer Testresultater
Appearance | Colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...