Salicylnitril ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: o-Cyanophenol, o-Hydroxybenzonitrile, 2-Cyanophenol, 2-Hydroxybenzonitrile

C1045-25G C1045-5G
TCIAC1045-25GEA 526 DKK
TCIAC1045-25G TCIAC1045-5G
Salicylnitril ≥98.0% (by GC, titration analysis)
Salicylnitril
Formel: C₇H₅NO
Kogepunkt: 149°C
Smeltepunkt: 90…92 °C
Massefylde: 1,1052 g/cm³ (99.6 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00002145
CAS-nummer: 611-20-1
EINECS: 210-259-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=601f0ff3b5677d946e735832dad3785a8dfec3c5&vid=2dbc4995ee31b911e2b99821076ed14fb734c325&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light red powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 94.0 to 99.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR