4-Aminobenzonitril ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
A0695-10G
A0695-250G
TCIAA0695-10GEA
337
DKK
TCIAA0695-10G
TCIAA0695-250G
4-Aminobenzonitril ≥98.0% (by GC, titration analysis)
4-Aminobenzonitril
Formel:
C₇H₆N₂ Kogepunkt: 167°C Smeltepunkt: 84…87 °C Massefylde: 1,274 g/cm³ (20 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00007821 CAS-nummer: 873-74-5 EINECS: 212-850-1 UN nummer: 3439 ADR: 6.1,III |
|
Specifikationer Testresultater
Appearance | White to Light yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 84.0 to 89.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...