3-Aminobenzonitril ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

A1203-25G A1203-5G
TCIAA1203-25GEA 1100 DKK
TCIAA1203-25G TCIAA1203-5G
3-Aminobenzonitril ≥98.0% (by GC, titration analysis)
3-Aminobenzonitril
Formel: C₇H₆N₂
Kogepunkt: 290°C
Smeltepunkt: 48…50 °C
Massefylde: 1,14 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007756
CAS-nummer: 2237-30-1
EINECS: 218-800-5
UN nummer: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0ab98d31a38b8fd321df79cd89cdbbe1276a29f&vid=53a36d861d523f6f29c25eeaab939fb4f2312f67&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light gray to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 52.0 to 55.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR