3-Aminobenzonitril ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
A0860-25G
A0860-250G
A0860-5G
TCIAA0860-25GEA
985
DKK
TCIAA0860-25G
TCIAA0860-250G
TCIAA0860-5G
3-Aminobenzonitril ≥98.0% (by GC, titration analysis)
3-Aminobenzonitril
Formel:
C₇H₆N₂ Kogepunkt: 290°C Smeltepunkt: 48…50 °C Massefylde: 1,14 g/cm³ (20 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00007756 CAS-nummer: 2237-30-1 EINECS: 218-800-5 UN nummer: 3439 ADR: 6.1,III |
|
Specifikationer Testresultater
Appearance | White to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 51.0 to 55.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...