2,4-Dimethoxybenzaldehyd ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

D0626-25G D0626-500G D0626-100G
TCIAD0626-25GEA 550 DKK
TCIAD0626-25G TCIAD0626-500G TCIAD0626-100G
2,4-Dimethoxybenzaldehyd ≥98.0% (by GC)
2,4-Dimethoxybenzaldehyde
Formel: C₉H₁₀O₃
Kogepunkt: 308 °C (1013 hPa)
Smeltepunkt: 67…68 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003311
CAS-nummer: 613-45-6
EINECS: 210-342-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=586f648ddb467d302b7ac3ff3984a8e4f41a3f43&vid=2c4e1e5c40d48b688dccb42b76e0e5355a1e33aa&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 69.0 to 72.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR