2-Hydroxy-4-methoxybenzaldehyd ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Hydroxy-4-methoxybenzaldehyde

H0699-25G H0699-5G
TCIAH0699-25GEA 1260 DKK
TCIAH0699-25G TCIAH0699-5G
2-Hydroxy-4-methoxybenzaldehyd ≥98.0% (by GC, titration analysis)
4-Methoxysalicylaldehyde
Formel: C₈H₈O₃
Kogepunkt: 124°C
Smeltepunkt: 42…43 °C
Massefylde: 1,2065 g/cm³ (51 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003327
CAS-nummer: 673-22-3
EINECS: 211-604-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6867971e1ab62d251f484c2b3a94e672630914a&vid=7aef82a984d63e77263c6ce2a01f9c34416413c2&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light red powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 40.0 to 44.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR