3-Nitrobenzaldehyd ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

N0129-25G N0129-500G
TCIAN0129-25GEA 140 DKK
TCIAN0129-25G TCIAN0129-500G
3-Nitrobenzaldehyd ≥98.0% (by GC)
3-Nitrobenzaldehyde
Formel: C₇H₅NO₃
Molekylvægt: 151.12 g/mol
Kogepunkt: 114°C
Smeltepunkt: 58
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007249
CAS-nummer: 99-61-6
EINECS: 202-772-6
Merck Index: 12,06684
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e640551dfa70207e0d17b6246d1ce5215b6d1477&vid=e5dff4df3ef7a5e62367519f7ab53ec402a9a829&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 55.0 to 59.0 deg-C
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR