2-Bromphenol ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
B0630-25G
B0630-500G
B0630-100G
TCIAB0630-25GEA
378
DKK
TCIAB0630-25G
TCIAB0630-500G
TCIAB0630-100G
2-Bromphenol ≥98.0% (by GC, titration analysis)
2-Bromphenol
Formel:
C₆H₅BrO Kogepunkt: 195 °C (1013 hPa) Massefylde: 1,49 g/cm³ (20 °C) Flammepunkt: 42°C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00002146 CAS-nummer: 95-56-7 EINECS: 202-432-7 UN nummer: 2929 ADR: 6.1,II Merck Index: 14,01428 |
|
Specifikationer Testresultater
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...