2-Brom-4-nitroanisol ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
2-Bromo-1-methoxy-4-nitrobenzene
B2830-25G
B2830-5G
TCIAB2830-25GEA
2540
DKK
TCIAB2830-25G
TCIAB2830-5G
2-Brom-4-nitroanisol ≥98.0% (by GC)
2-Brom-4-nitroanisol
Formel:
C₇H₆BrNO₃ |
MDL Number:
MFCD00041242 CAS-nummer: 5197-28-4 EINECS: 225-983-5 |
|
Specifikationer Testresultater
Appearance | White to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 102.0 to 106.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...