2-Brom-5-nitroanisol ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
B3418-25G
B3418-5G
TCIAB3418-25GEA
2060
DKK
TCIAB3418-25G
TCIAB3418-5G
2-Brom-5-nitroanisol ≥98.0% (by GC)
2-Brom-5-nitroanisol
Formel:
C₇H₆BrNO₃ |
MDL Number:
MFCD00041250 CAS-nummer: 77337-82-7 EINECS: 278-669-5 |
|
Specifikationer Testresultater
Appearance | Light yellow to Brown to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 102.0 to 106.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...