1-Brom-3-nitrobenzen ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
B0623-25G
B0623-500G
B0623-100G
TCIAB0623-25GEA
230
DKK
TCIAB0623-25G
TCIAB0623-500G
TCIAB0623-100G
1-Brom-3-nitrobenzen ≥98.0% (by GC)
1-Brom-3-nitrobenzen
Formel:
C₆H₄BrNO₂ Kogepunkt: 256 °C (1013 hPa) Smeltepunkt: 48…51 °C Massefylde: 1,969 g/cm³ (20 °C) Flammepunkt: (closed cup) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00024298 CAS-nummer: 585-79-5 EINECS: 209-563-9 UN nummer: 3459 ADR: 6.1,III |
|
Specifikationer Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 54.0 to 56.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...