2-Brom-5-nitroanilin ≥97.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
B3041-25G
B3041-5G
TCIAB3041-25GEA
1640
DKK
TCIAB3041-25G
TCIAB3041-5G
2-Brom-5-nitroanilin ≥97.0% (by GC)
2-Brom-5-nitroanilin
Formel:
C₆H₅BrN₂O₂ Molekylvægt: 217.02 g/mol Smeltepunkt: 141 |
MDL Number:
MFCD00051578 CAS-nummer: 10403-47-1 EINECS: 233-874-9 |
|
Specifikationer Testresultater
Appearance | White to Brown to Dark purple powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 138.0 to 142.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...