3-Bromanilin ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B0542-25G B0542-100G
TCIAB0542-25GEA 312 DKK
TCIAB0542-25G TCIAB0542-100G
3-Bromanilin ≥98.0% (by GC)
3-Bromanilin
Formel: C₆H₆BrN
Kogepunkt: 251°C
Smeltepunkt: 16 °C
Massefylde: 1,59 g/cm³ (20 °C)
Flammepunkt: ≥ 230 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007757
CAS-nummer: 591-19-5
EINECS: 209-704-4
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ad4bafd76eff30d86621241f0af3928acd41da79&vid=ad4bafd76eff30d86621241f0af3928acd41da79&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White or Colorless to Light orange to Yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR