2,5-Dibromanilin ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

D1989-25G D1989-5G
TCIAD1989-25GEA 747 DKK
TCIAD1989-25G TCIAD1989-5G
2,5-Dibromanilin ≥98.0% (by GC)
2,5-Dibromanilin
Formel: C₆H₅Br₂N
Kogepunkt: 281 °C (1013 hPa)
MDL Number: MFCD00007636
CAS-nummer: 3638-73-1
EINECS: 222-865-5
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b4adaa6f767e098669fb14608f0b62ee5297a7d1&vid=3ef4676824bb19b6a4d1cd104823c8c039117c24&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 54.0 to 58.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR