4-Brom-2-fluorbenzoesyre ≥99.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B1950-25G B1950-5G
TCIAB1950-25GEA 1260 DKK
TCIAB1950-25G TCIAB1950-5G
4-Brom-2-fluorbenzoesyre ≥99.0% (by titrimetric analysis)
4-Brom-2-fluorbenzoesyre
Formel: C₇H₄BrFO₂
MDL Number: MFCD00143263
CAS-nummer: 112704-79-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba12066c03261859ea4a9258f8137da4a519d2ae&vid=7c7f440d808d8bf0ad4d3c6ed7dba1a33dcc9aaf&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) min. 99.0 %
Melting point 208.0 to 213.0 deg-C
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR