3-Brom-4-Fluorbenzonitril ≥99.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
B1965-25G
B1965-5G
TCIAB1965-25GEA
673
DKK
TCIAB1965-25G
TCIAB1965-5G
3-Brom-4-Fluorbenzonitril ≥99.0% (by GC)
3-Brom-4-Fluorbenzonitril
Formel:
C₇H₃BrFN Kogepunkt: 134 - 136 °C (33 -rr) Smeltepunkt: 53…58 °C |
MDL Number:
MFCD00055432 CAS-nummer: 79630-23-2 EINECS: 279-200-7 UN nummer: 3439 ADR: 6.1,III |
|
Specifikationer Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 55.0 to 58.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...