4-fluorphenylhydraziniumchlorid ≥97.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: (4-Fluorphenyl)hydrazin HCl, 4-Fluorophenylhydrazine hydrochloride, 1-(4-Fluorophenyl)hydrazine hydrochloride, (4-Fluorophenyl)hydrazine hydrochloride

F0246-25G F0246-5G
TCIAF0246-25GEA 1540 DKK
TCIAF0246-25G TCIAF0246-5G
4-fluorphenylhydraziniumchlorid ≥97.0% (by HPLC, titration analysis)
4-fluorphenylhydraziniumchlorid
Formel: C₆H₈ClFN₂
MDL Number: MFCD00012942
CAS-nummer: 823-85-8
EINECS: 212-521-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2adfadf2cee302c3e773f05d6afbf6497fcd89ee&vid=2adfadf2cee302c3e773f05d6afbf6497fcd89ee&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Potassium iodate Method) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR