2,3-Dichlorphenylhydraziniumchlorid ≥97.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
D2641-25G
D2641-5G
TCIAD2641-25GEA
5350
DKK
TCIAD2641-25G
TCIAD2641-5G
2,3-Dichlorphenylhydraziniumchlorid ≥97.0% (by titrimetric analysis)
2,3-Dichlorphenylhydraziniumchlorid
Formel:
C₆H₇Cl₃N₂ |
MDL Number:
MFCD00052265 CAS-nummer: 21938-47-6 |
|
Specifikationer Testresultater
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (Potassium iodate Method) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...