3-Bromphenylhydraziniumchlorid ≥98.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B3847-25G B3847-5G
TCIAB3847-25GEA 1240 DKK
TCIAB3847-25G TCIAB3847-5G
3-Bromphenylhydraziniumchlorid ≥98.0% (by HPLC)
3-Bromphenylhydraziniumchlorid
Formel: C₆H₈BrClN₂
Smeltepunkt: 225°C
MDL Number: MFCD00012933
CAS-nummer: 27246-81-7
EINECS: 248-362-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e28d1cf177988ba9e84a16181f14bf2923e13a98&vid=e28d1cf177988ba9e84a16181f14bf2923e13a98&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Potassium iodate Method) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR