Allyl trifluoracetat ≥95.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Allyl Trifluoroacetate, ['Trifluoroacetic acid Allyl Ester'], Trifluorethansäure(2-propenyl)ester, Trifluoroacetic allyl ester

T1765-25G T1765-5G
TCIAT1765-25GEA 1350 DKK
TCIAT1765-25G TCIAT1765-5G
Allyl trifluoracetat ≥95.0% (by GC)
Allyl trifluoracetat
Formel: C₅H₅F₃O₂
Molekylvægt: 154.09 g/mol
Kogepunkt: 66°C
Flammepunkt: –1°C
MDL Number: MFCD00013567
CAS-nummer: 383-67-5
EINECS: 206-853-7
UN nummer: 2924
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ae77c55516e78af1ff892beba37e3a603ff9b72&vid=0ae77c55516e78af1ff892beba37e3a603ff9b72&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR