di-Natriumhydrogencitrat sesquihydrat ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Citric acid disodium salt sesquihydrate, Sodium hydrogencitrate sesquihydrate, Disodium hydrogen 2-hydroxypropane-1,2,3-tricarboxylate sesquihydrate, Sodium citrate dibasic sesquihydrate
C0365-25G
C0365-500G
TCIAC0365-25GEA
140
DKK
TCIAC0365-25G
TCIAC0365-500G
di-Natriumhydrogencitrat sesquihydrat ≥98.0% (by titrimetric analysis)
di-Natriumhydrogencitrat sesquihydrat
Formel:
C₆H₆Na₂O₇·1.5H₂O Smeltepunkt: 149°C |
MDL Number:
MFCD00150445 CAS-nummer: 6132-05-4 EINECS: 205-623-3 Merck Index: 14,02326 |
|
Specifikationer Testresultater
Appearance | White powder to crystaline |
Purity (Nonaqueous Titration) | min. 98.0 % (calcd.on anh.substance) |
Solubility in Water | almost transparency |
Water | 9.0 to 12.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...