4-Brom-1,2-difluorbenzen ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1-Brom-3,4-difluorbenzen

B1523-25G B1523-250G
TCIAB1523-25GEA 296 DKK
TCIAB1523-25G TCIAB1523-250G
4-Brom-1,2-difluorbenzen ≥98.0% (by GC)
4-Brom-1,2-difluorbenzen
Formel: C₆H₃BrF₂
Molekylvægt: 192.99 g/mol
Kogepunkt: 151°C
Smeltepunkt: -4
Massefylde: 1.7
Flammepunkt: 33°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00000304
CAS-nummer: 348-61-8
EINECS: 206-481-5
UN nummer: 1993
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=921509fa0d7ed1d60bde8dcffcc1cc26fe9052a8&vid=b40cfb305f91929a7120b701712a5d33187aa12d&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR