1-Brom-3,4,5-trifluorbenzen ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 5-Brom-1,2,3-trifluorbenzen

B1765-25G B1765-5G
TCIAB1765-25GEA 1010 DKK
TCIAB1765-25G TCIAB1765-5G
1-Brom-3,4,5-trifluorbenzen ≥98.0% (by GC)
1-Brom-3,4,5-trifluorbenzen
Formel: C₆H₂BrF₃
Kogepunkt: 43°C
Smeltepunkt: –20 °C
Massefylde: 1,767 g/cm³ (20 °C)
Flammepunkt: 45°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00070738
CAS-nummer: 138526-69-9
EINECS: 418-480-9
UN nummer: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d4db6094cdccf983d35899b87c72e6ab97ef5d4&vid=caf1b78a77d0cdc567df07a69fd14d4238185fd7&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR