1-Brom-2,3,5,6-tetrafluorbenzen ≥97.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
2,3,5,6-Tetrafluorobromobenzene, 3-Bromo-1,2,4,5-tetrafluorobenzene
B1706-25G
B1706-5G
TCIAB1706-25GEA
2630
DKK
TCIAB1706-25G
TCIAB1706-5G
1-Brom-2,3,5,6-tetrafluorbenzen ≥97.0% (by GC)
1-Brom-2,3,5,6-tetrafluorbenzen
Formel:
C₆HBrF₄ Kogepunkt: 143°C Massefylde: 1,88 g/cm³ (20 °C) Flammepunkt: 54°C |
MDL Number:
MFCD00000308 CAS-nummer: 1559-88-2 EINECS: 216-327-9 UN nummer: 1993 ADR: 3,III |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...