1,4-Dibrom-2,5-difluorbenzen ≥98.0% (by GC)
Leverandør: TCI
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
D1864-10G D1864-25G
TCIAD1864-10GEA 378 DKK
TCIAD1864-10G TCIAD1864-25G
1,4-Dibrom-2,5-difluorbenzen ≥98.0% (by GC)
1,4-Dibrom-2,5-difluorbenzen
Formel: C₆H₂Br₂F₂
Molekylvægt: 271.89 g/mol
Kogepunkt: 96°C
Smeltepunkt: 65
MDL Number: MFCD00000346
CAS-nummer: 327-51-5
EINECS: 206-317-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6953d457c66a6de9c766bd26ae2ffa3be6de2c9&vid=c6114ced5fc8f88ff150ac3057e9b8466fd14fe2&prid=web&clid=SAPDAM
Bestil nu
Specifikationer Testresultater
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 62.0 to 66.0 deg-C
Solubility in Methanol almost transparency