2-Brom-5-chlorfluorbenzen ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B2724-5G B2724-25G
TCIAB2724-5GEA 181 DKK
TCIAB2724-5G TCIAB2724-25G
2-Brom-5-chlorfluorbenzen ≥98.0% (by GC)
2-Brom-5-chlorfluorbenzen
Formel: C₆H₃BrClF
Kogepunkt: 92°C
Flammepunkt: 92°C
MDL Number: MFCD00079708
CAS-nummer: 1996-29-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b8b4210fb425950e23c14cc1a11b671407dc3a9&vid=7b8b4210fb425950e23c14cc1a11b671407dc3a9&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR