4-Brom-2-chlorfluorbenzen ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 4-Bromo-2-chloro-1-fluorobenzene, 4-Bromo-2-chlorofluorobenzene

B1860-25G B1860-5G
TCIAB1860-25GEA 698 DKK
TCIAB1860-25G TCIAB1860-5G
4-Brom-2-chlorfluorbenzen ≥98.0% (by GC)
4-Brom-2-chlorfluorbenzen
Formel: C₆H₃BrClF
Kogepunkt: 95°C
Massefylde: 1,72 g/cm³ (20 °C)
Flammepunkt: 88°C
MDL Number: MFCD00051794
CAS-nummer: 60811-21-4
EINECS: 405-580-2
UN nummer: 3082
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8167bd867b54775c84b131652732dab163f8dd7a&vid=b12681de55dbacb300f7c180a3095246a1575e2a&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR