4-Brom-1,2-dichlorbenzen ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1-Bromo-3,4-dichlorobenzene

B0983-25G B0983-500G B0983-100G
TCIAB0983-25GEA 378 DKK
TCIAB0983-25G TCIAB0983-500G TCIAB0983-100G
4-Brom-1,2-dichlorbenzen ≥98.0% (by GC)
4-Brom-1,2-dichlorbenzen
Formel: C₆H₃BrCl₂
Molekylvægt: 225.9 g/mol
Kogepunkt: 140
Smeltepunkt: 24.5
MDL Number: MFCD00040849
CAS-nummer: 18282-59-2
EINECS: 242-160-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=453d4dce7d0a49ab4d968e33e72dac67c5e524c6&vid=1429d58aadb86e97b9573b3c495652ad1edf02a2&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White or Colorless to Light orange to Yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR